missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ D4 Dopamine Receptor Recombinant Rabbit Monoclonal Antibody (HL2851)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556685
This item is not returnable.
View return policy
Description
D4 Dopamine Receptor Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes the D4 subtype of the dopamine receptor. The D4 subtype is a G-protein coupled receptor which inhibits adenylyl cyclase. It is a target for drugs which treat schizophrenia and Parkinson disease. Mutations in this gene have been associated with various behavioral phenotypes, including autonomic nervous system dysfunction, attention deficit/hyperactivity disorder, and the personality trait of novelty seeking. This gene contains a polymorphic number (2-10 copies) of tandem 48 nt repeats; the sequence shown contains four repeats.
Specifications
| D4 Dopamine Receptor | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P21917 | |
| DRD4 | |
| Synthetic peptide encompassing a sequence within the Extracellular domain human of Dopamine Receptor D4. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| HL2851 | |
| Unconjugated | |
| DRD4 | |
| AW125663; D(2C) dopamine receptor; D(4) dopamine receptor; D4DR; D4R; D4RA; Dopamine 4 receptor; Dopamine D4 receptor; dopamine receptor 4; dopamine receptor D4; DRD4; Drd-4; seven transmembrane helix receptor | |
| Rabbit | |
| Protein A | |
| RUO | |
| 1815 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction