missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DAAM2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
222.00 € - 487.00 €
Specifications
| Antigen | DAAM2 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18676702
|
Novus Biologicals
NBP2-92809-0.02ml |
0.02 mL |
222.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18600662
|
Novus Biologicals
NBP2-92809-0.1ml |
0.1 mL |
487.00 €
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DAAM2 Polyclonal antibody specifically detects DAAM2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| DAAM2 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Stem Cells | |
| PBS with 50% glycerol, pH7.3. | |
| 23500 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| dishevelled associated activator of morphogenesis 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-86 of human DAAM2 (NP_056160.2). MAPRKRSHHGLGFLCCFGGSDIPEINLRDNHPLQFMEFSSPIPNAEELNIRFAELVDELDLTDKNREAMFALPPEKKWQIYCSKKK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title