missing translation for 'onlineSavingsMsg'
Learn More

Dabrafenib, MedChemExpress

Product Code. 16416128
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16416128 10 mg 10mg
16482257 50 mg 50mg
16468727 5 mg 5mg
16451136 200 mg 200mg
15750389 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16416128 Supplier Medchem Express Supplier No. HY14660
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Dabrafenib (GSK2118436A) is an ATP-competitive inhibitor of Raf with IC50s of 5 nM and 0.6 nM for C-Raf and B-RafV600E, respectively.

Dabrafenib (GSK2118436A) is an ATP-competitive inhibitor of Raf with IC50s of 5 nM and 0.6 nM for C-Raf and B-RafV600E, respectively.

Specifications

Chemical Name or Material Dabrafenib
CAS 1195765-45-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C23H20F3N5O2S2
Quantity 10 mg
Synonym GSK2118436A GSK2118436
Solubility Information DMSO : ≥ 33 mg/mL (63.52 mM)
SMILES CC(C)(C)C1=NC(C2=C(F)C(NS(C3=C(F)C=CC=C3F)(=O)=O)=CC=C2)=C(C4=CC=NC(N)=N4)S1
Molecular Weight (g/mol) 519.56
Formula Weight 519.56
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.