missing translation for 'onlineSavingsMsg'
Learn More

Dabrafenib Mesylate, TRC

Product Code. 30138620
Change view
Click to view available options
Quantity:
10 mg
500 mg
Unit Size:
10mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138620 500 mg 500mg
17971945 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138620 Supplier Toronto Research Chemicals Supplier No. TRCD101005500MG
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1195768-06-9
Molecular Formula C23 H20 F3 N5 O2 S2 . C H4 O3 S
Molecular Weight (g/mol) 615.67
Synonym Benzenesulfonamide, N-[3-[5-(2-amino-4-pyrimidinyl)-2-(1,1-dimethylethyl)-4-thiazolyl]-2-fluorophenyl]-2,6-difluoro-, methanesulfonate (1:1), Dabrafenib mesylate, GSK 2118436 methanesulfonate salt, GSK 2118436B, N-[3-[5-(2-Aminopyrimidin-4-yl)-2-tert-butyl-1,3-thiazol-4-yl]-2-fluorophenyl]-2,6-difluorobenzenesulfonamide methanesulfonic acid, Benzenesulfonamide, N-[3-[5-(2-amino-4-pyrimidinyl)-2-(1,1-dimethylethyl)-4-thiazolyl]-2-fluorophenyl]-2,6-difluoro-, methanesulfonate (1:1), Dabrafenib mesylate, GSK 2118436 methanesulfonate salt, GSK 2118436B, N-[3-[5-(2-Aminopyrimidin-4-yl)-2-tert-butyl-1,3-thiazol-4-yl]-2-fluorophenyl]-2,6-difluorobenzenesulfonamide methanesulfonic acid, GSK 2118436B
IUPAC Name N-[3-[5-(2-aminopyrimidin-4-yl)-2-tert-butyl-1,3-thiazol-4-yl]-2-fluorophenyl]-2,6-difluorobenzenesulfonamide;methanesulfonic acid
SMILES CC(C)(C)c1nc(c2cccc(NS(=O)(=O)c3c(F)cccc3F)c2F)c(s1)c4ccnc(N)n4.CS(=O)(=O)O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C23H20F3N5O2S2.CH4O3S/c1-23(2,3)21-30-18(19(34-21)16-10-11-28-22(27)29-16)12-6-4-9-15(17(12)26)31-35(32,33)20-13(24)7-5-8-14(20)25;1-5(2,3)4/h4-11,31H,1-3H3,(H2,27,28,29);1H3,(H,2,3,4)
Formula Weight 615.0892
Chemical Name or Material Dabrafenib Mesylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.