missing translation for 'onlineSavingsMsg'
Learn More

Dacarbazine, MedChemExpress

Product Code. 16451366
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mg
1 g
Unit Size:
1g
200mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16451366 200 mg 200mg
16402049 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16451366 Supplier Medchem Express Supplier No. HYB0078200mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Dacarbazine is a cell cycle nonspecific antineoplastic alkylating agent. Dacarbazine inhibits T and B lymphoblastic response, with IC50 values of 50 and 10 μg/ml, respectively. Dacarbazine can be used for the research of metastatic malignant melanoma.

Dacarbazine is a cell cycle nonspecific antineoplastic alkylating agent. Dacarbazine inhibits T and B lymphoblastic response, with IC50 values of 50 and 10 μg/ml, respectively. Dacarbazine can be used for the research of metastatic malignant melanoma.

Specifications

Chemical Name or Material Dacarbazine
Physical Form Solid
CAS 3-4-4342
Color White
Quantity 200 mg
Purity Grade Notes Research
Molecular Formula C6H10N6O
For Use With (Application) Cancer-programmed cell death
Synonym Imidazole Carboxamide
Solubility Information DMSO : 5 mg/mL (27.45 mM; ultrasonic and warming and heat to 60°C) ∣H2O : 1 mg/mL (5.49 mM; Need ultrasonic and warming)
SMILES O=C(C1=C(/N=N/N(C)C)NC=N1)N
Molecular Weight (g/mol) 182.18
Formula Weight 182.18
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.