missing translation for 'onlineSavingsMsg'
Learn More

Daidzein, 98.7%, MP Biomedicals

Product Code. 11459981 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
20 mg
100 mg
Unit Size:
100mg
20mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11459981 100 mg 100mg
11449981 20 mg 20mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11459981 Supplier MP Biomedicals Supplier No. 0215881280
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 486-66-8
Molecular Formula C15H10O4
Molecular Weight (g/mol) 254.241
InChI Key ZQSIJRDFPHDXIC-UHFFFAOYSA-N
Synonym daidzein, 4',7-dihydroxyisoflavone, daidzeol, 7,4'-dihydroxyisoflavone, 7-hydroxy-3-4-hydroxyphenyl-4h-chromen-4-one, diadzein, 7-hydroxy-3-4-hydroxyphenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 7-hydroxy-3-4-hydroxyphenyl, 7-hydroxy-3-4-hydroxyphenyl chromen-4-one, 7-hydroxy-3-4-hydroxyphenyl-4-benzopyrone
PubChem CID 5281708
ChEBI CHEBI:28197
IUPAC Name 7-hydroxy-3-(4-hydroxyphenyl)chromen-4-one
SMILES C1=CC(=CC=C1C2=COC3=C(C2=O)C=CC(=C3)O)O

Specifications

Color White
Assay Percent Range 0.987
Quantity 100 mg
Formula Weight 254.23
Physical Form Powder
Chemical Name or Material Daidzein, For HPLC analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.