missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Daidzin, MedChemExpress
Description
Daidzin is an isoflavone that has anti-oxidant, anti-carcinogenic, and anti-atherosclerotic activities
Specifications
Specifications
| Chemical Name or Material | Daidzin |
| CAS | 552-66-9 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C21H20O9 |
| Quantity | 10 mg |
| Synonym | Daidzoside NPI-031D Daidzein 7-O-glucoside |
| Solubility Information | DMSO : ≥ 34 mg/mL (81.66 mM) |
| SMILES | OC1=CC=C(C2=COC(C=C(O[C@H]3[C@H](O)[C@@H](O)[C@H](O)[C@@H](CO)O3)C=C4)=C4C2=O)C=C1 |
| Molecular Weight (g/mol) | 416.38 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?