missing translation for 'onlineSavingsMsg'
Learn More

Daidzin, MedChemExpress

Product Code. 16492336
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16492336 500 mg 500mg
16491476 200 mg 200mg
16474727 50 mg 50mg
16428828 10 mg 10mg
16468956 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16492336 Supplier Medchem Express Supplier No. HYN0018
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Daidzin is an isoflavone that has anti-oxidant, anti-carcinogenic, and anti-atherosclerotic activities; directly inhibits mitochondrial aldehyde dehydrogenase 2 (IC50 = 80 nM) and is an effective anti-dipsotropic isoflavone.

Daidzin is an isoflavone that has anti-oxidant, anti-carcinogenic, and anti-atherosclerotic activities

Specifications

Chemical Name or Material Daidzin
CAS 552-66-9
Color White
Purity Grade Notes Research
Molecular Formula C21H20O9
Quantity 500 mg
Synonym Daidzoside NPI-031D Daidzein 7-O-glucoside
Solubility Information DMSO : ≥ 34 mg/mL (81.66 mM)
SMILES OC1=CC=C(C2=COC(C=C(O[C@H]3[C@H](O)[C@@H](O)[C@H](O)[C@@H](CO)O3)C=C4)=C4C2=O)C=C1
Molecular Weight (g/mol) 416.38
Formula Weight 416.38
Percent Purity 98.39%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.