missing translation for 'onlineSavingsMsg'
Learn More

Danshensu sodium salt, MedChemExpress

Product Code. 16414757
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
This item is not returnable. View return policy

Product Code. 16414757

Brand: Medchem Express HYN0106

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Danshensu (sodium salt) is odium salt of danshensu from the widely used Chinese herb Danshen.

Danshensu (sodium salt) is odium salt of danshensu from the widely used Chinese herb Danshen.

Specifications

Chemical Name or Material Danshensu sodium salt
CAS 67920-52-9
Color White
Purity Grade Notes Research
Molecular Formula C9H9NaO5
Quantity 50 mg
Synonym Sodium Danshensu (±)-DanShenSu sodium sal
Solubility Information H2O : 100 mg/mL (454.24 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (454.24 mM; Need ultrasonic)
SMILES OC1=C(O)C=CC(CC(C(O[Na])=O)O)=C1
Molecular Weight (g/mol) 220.15
Formula Weight 220.15
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.