missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DAP3 Antibody (42C617.1.2) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB100-56539JF669
This item is not returnable.
View return policy
Description
DAP3 Monoclonal antibody specifically detects DAP3 in Human,Mouse samples. It is validated for Western Blot
Specifications
| DAP3 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| bMRP-10, DAP-3Death-associated protein 3, death associated protein 3, DKFZp686G12159,28S ribosomal protein S29, mitochondrial, FLJ12817, MGC126058, MGC126059, mitochondrial 28S ribosomal protein S29, MRP-S29Ionizing radiation resistance conferring protein, MRPS29S29mt | |
| The antibody was raised against a synthetic peptide corresponding to amino acids 51-66 of human DAP-3 protein. | |
| 0.1 mL | |
| Apoptosis, Core ESC Like Genes, Stem Cell Markers | |
| 7818 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| 42C617.1.2 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction