missing translation for 'onlineSavingsMsg'
Learn More

Dapsone, TRC

Product Code. 30137498
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
100g
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137498 100 g 100g
17950945 1 g 1g
17981535 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137498 Supplier Toronto Research Chemicals Supplier No. TRCD193250100G
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-08-0
Molecular Formula C12 H12 N2 O2 S
Molecular Weight (g/mol) 248.3
Synonym Aniline, 4,4'-sulfonyldi- (7CI,8CI), 4,4'-Sulfonylbis[benzenamine], 1,1'-Sulfonylbis[4-aminobenzene], 1358F, 4,4'-DDS, 4,4'-Dapsone, 4,4'-Diaminobiphenyl sulfone, 4,4'-Diaminodiphenyl sulfone, 4,4'-Sulfonylbisaniline, 4,4'-Sulfonylbisbenzamine, 4,4'-Sulfonyldianiline, 4,4'-Sulfonyldiphenylamine, 4,4'-Sulphonyldianiline, 4-(4-Aminophenyl)sulfonylaniline, 4-(4-Aminophenylsulfonyl)benzenamine, 4-Aminophenyl sulfone, Aczone, Aradur 9664-1, Aradur 9664-1NL, Aradur 976, Aradur 976-1, Aradur 9761-1, Araldite HT 976, Atrisone, Avlosulfon, Avlosulfone, Bis(4-aminophenyl) sulfone, Bis(p-aminophenyl) sulfone, Croysulfone, D 0089, DADFS, DADPS, DDS, DDS (pharmaceutical), DSS, Dapson, Dapsone, Di(4-aminophenyl)sulfone, Di(p-aminophenyl) sulfone, Diaphenylsulfon, Diaphenylsulfone, Diphenasone, Diphone, Disulone, Dumitone, Eporal, F 1358, HB-MC-GE 40, HB-MC-GE 45, HEW 62, HT 9664, HT 976, HT 976-1, HY 976, Hardener HT 976, Lapox K 10, Metabolite C, NSC 6091, Novophone, Seikacure S, Servidapson, Sulfona, Sulfona-Mae, Sulphadione, Sumicure S, Udolac, p,p'-Diaminodiphenyl sulfone, p,p'-Sulfonyldianiline, p-Aminophenyl sulfone
IUPAC Name 4-(4-aminophenyl)sulfonylaniline
SMILES Nc1ccc(cc1)S(=O)(=O)c2ccc(N)cc2

Specifications

Quantity 100 g
InChI Formula InChI=1S/C12H12N2O2S/c13-9-1-5-11(6-2-9)17(15,16)12-7-3-10(14)4-8-12/h1-8H,13-14H2
Formula Weight 248.0619
Percent Purity >95%
Chemical Name or Material Dapsone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.