missing translation for 'onlineSavingsMsg'
Learn More

Daptomycin, MedChemExpress

Product Code. 16454237
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16454237 50 mg 50mg
16498096 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16454237 Supplier Medchem Express Supplier No. HYB0108
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Daptomycin is a lipopeptide antibiotic with rapid in vitro bactericidal activity against gram-positive organisms.

Daptomycin is a lipopeptide antibiotic with rapid in vitro bactericidal activity against gram-positive organisms.

Specifications

Chemical Name or Material Daptomycin
Physical Form Powder
CAS 103060-53-3
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C72H101N17O26
For Use With (Application) COVID-19-immunoregulation
Synonym LY146032
Solubility Information H2O : 100 mg/mL (61.70 mM; Need ultrasonic) ∣DMSO : ≥ 100 mg/mL (61.70 mM)
SMILES O=C(N[C@H](CC(N)=O)C(N[C@@H](CC(O)=O)C(N[C@H](C(NCC(N[C@H](C(N[C@@](C(N[C@@H](C(N[C@H]1CC(O)=O)=O)C)=O)([H])CC(O)=O)=O)CCCN)=O)=O)[C@H](OC([C@](NC([C@](NC([C@@](NC(CNC1=O)=O)([H])CO)=O)([H])[C@H](C)CC(O)=O)=O)([H])CC(C(C=CC=C2)=C2N)=O)=O)C)=O)=O)[C@@H](NC(CCCCCCCCC)=O)CC3=CNC4=CC=CC=C34
Molecular Weight (g/mol) 1620.67
Formula Weight 1620.67
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.