missing translation for 'onlineSavingsMsg'
Learn More

TOKU-E™ Daptomycin ReadyMade™ Solution

Product Code. 30044059
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 0.5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30044059 10 x 0.5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30044059 Supplier TOKU-E™ Supplier No. D10810x0.5ML
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Daptomycin ReadyMade™ Solution is provided as a sterile-filtered solution in ~ 10% β-cyclodextrin saline solution.

  • CAS Number: 103060-53-3
  • Molecular Formula: C72H101N17O26
  • Molecular Weight: 1620.67 g/mol
  • Storage Conditions: -20°C
  • Form: Liquid
  • Source: Streptomyces roseosporus
  • Spectrum: Daptomycin targets Gram-positive organisms including GRE (glycopeptide-resistant Enterococci) and MRSA (methicillin-resistant Staphylococcus aureus)
  • Mechanism of Action: Daptomycin peptides interact with anionic phospholipids in the membrane of Gram-positive bacteria. Peptides integrate into the membrane by a calcium-mediated process and interact with membrane phospholipid head groups and acyl chains. Finally, a pore or channel is formed which can kill the cell by membrane disruption/depolarization or cellular uptake of Daptomycin leading to disruption of intracellular processes such as synthesis of DNA, RNA, and protein which results in bacterial cell death. The activity of Daptomycin is dependent on the presence of physiologic levels of free calcium ions

Specifications

Chemical Name or Material Daptomycin
Physical Form Liquid
CAS 103060-53-3
pH 6.5
Source Streptomyces roseosporus
Packaging 2 mL plastic vial
Sterile Filtered Yes, 0.22 μm
Sterility Sterile
Color Clear Colorless to Pale Yellow
Concentration 1 to 1.05 mg/mL
Quantity 10 x 0.5 mL
Purity Grade Notes >95% (HPLC)
Assay Percent Range >95.0%
Assay Percent Notes Daptomycin, Dried Basis
Molecular Formula C72H101N17O26
Product Line ReadyMade™ Solutions
Shipping Condition Ice Pack
Synonym LY-146032; Cubicin
IUPAC Name (3S)-3-[[(2R)-4-amino-2-[[(2S)-2-(decanoylamino)-3-(1H-indol-3-yl)propanoyl]amino]-4-oxobutanoyl]amino]-4-[[(3S,6S,9R,15S,18R,21S,24S,30S,31R)-3-[2-(2-aminophenyl)-2-oxoethyl]-24-(3-aminopropyl)-15,21-bis(carboxymethyl)-6-[(2R)-1-carboxypropan-2-yl]-9-(hydroxymethyl)-18,31-dimethyl-2,5,8,11,14,17,20,23,26,29-decaoxo-1-oxa-4,7,10,13,16,19,22,25,28-nonazacyclohentriacont-30-yl]amino]-4-oxobutanoic acid
Molecular Weight (g/mol) 1620.67 g/mol
Formula Weight 1620.67 amu
Percent Purity >95%
Grade Antibiotic
Concentration or Composition (by Analyte or Components) Daptomycin: 0.1%; Sulfobutyl Ether beta;-Cyclodextrin: 5 to 50%
Show More Show Less
Product Title
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.