missing translation for 'onlineSavingsMsg'
Learn More

Darifenacin hydrobromide, MedChemExpress

Product Code. p-200100757
Encompass_Preferred
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
This item is not returnable. View return policy

Product Code. 16488258

Brand: Medchem Express HYA0012

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Darifenacin hydrobromide (UK-88525 hydrobromide) is a selective M3 muscarinic receptor antagonist with pKi of 8.9.

Darifenacin hydrobromide (UK-88525 hydrobromide) is a selective M3 muscarinic receptor antagonist with pKi of 8.9.

Specifications

Chemical Name or Material Darifenacin hydrobromide
CAS 133099-07-7
Color White
Purity Grade Notes Research
Molecular Formula C28H31BrN2O2
Quantity 10 mg
Synonym UK-88525 hydrobromide
Solubility Information DMSO : 33.33 mg/mL (65.68 mM; Need ultrasonic)
SMILES O=C(C(C1=CC=CC=C1)([C@@H]2CCN(CCC3=CC4=C(OCC4)C=C3)C2)C5=CC=CC=C5)N.Br
Molecular Weight (g/mol) 507.46
Formula Weight 507.46
Percent Purity 95.04%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.