Learn More
DARS Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand:  Novus Biologicals NBP1-85937
Product Code. 18707693
  • 547.00 € / 0.10mL
Description
DARS Polyclonal specifically detects DARS in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockdown Validated.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.