missing translation for 'onlineSavingsMsg'
Learn More

Darunavir, MedChemExpress

Product Code. 16489797
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16489797 5 mg 5mg
16457288 10 mg 10mg
16423307 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16489797 Supplier Medchem Express Supplier No. HY17040
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Darunavir (TMC114), an orally active next generation HIV protease inhibitor, has a similar antiviral activity against the mutant and the wild-type viruses. Darunavir (TMC114) is potent against laboratory HIV-1 strains and primary clinical isolates (IC50 = 0.003 μM; IC90 = 0.009 μM) with minimal cytotoxicity.

Darunavir (TMC114), an orally active next generation HIV protease inhibitor, has a similar antiviral activity against the mutant and the wild-type viruses. Darunavir (TMC114) is potent against laboratory HIV-1 strains and primary clinical isolates (IC50 = 0.003 μM

Specifications

Chemical Name or Material Darunavir
Physical Form Solid
CAS 206361-99-1
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C27H37N3O7S
For Use With (Application) COVID-19-anti-virus
Synonym TMC114 UIC-94017
Solubility Information DMSO : ≥ 100 mg/mL (182.60 mM)
SMILES O=C(O[C@@H]1[C@@]2([H])[C@@](OCC2)([H])OC1)N[C@@H](CC3=CC=CC=C3)[C@H](O)CN(S(=O)(C4=CC=C(N)C=C4)=O)CC(C)C
Molecular Weight (g/mol) 547.66
Formula Weight 547.66
Percent Purity 99.84%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.