missing translation for 'onlineSavingsMsg'
Learn More

Daunorubicinol Hydrochloride (Mixture of Diastereomers), TRC

Product Code. 18081030
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
25 mg
Unit Size:
1mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18081030 25 mg 25mg
30133481 1 mg 1mg
18033330 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18081030 Supplier Toronto Research Chemicals Supplier No. TRCD19450525MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28008-53-9
Molecular Formula C27 H31 N O10 . Cl H
Molecular Weight (g/mol) 566

Specifications

Quantity 25 mg
Synonym 5,12-Naphthacenedione, 10-[(3-amino-2,3,6-trideoxy-α-L-lyxo-hexopyranosyl)oxy]-7,8,9,10-tetrahydro-6,8,11-trihydroxy-8-[(1S)-1-hydroxyethyl]-1-methoxy-, hydrochloride (1:1), (8S,10S)-, 5,12-Naphthacenedione, 10-[(3-amino-2,3,6-trideoxy-α-L-lyxo-hexopyranosyl)oxy]-7,8,9,10-tetrahydro-6,8,11-trihydroxy-8-(1-hydroxyethyl)-1-methoxy-, hydrochloride, [8S-[8α,8(R*),10α]]-, 5,12-Naphthacenedione, 10-[(3-amino-2,3,6-trideoxy-α-L-lyxo-hexopyranosyl)oxy]-7,8,9,10-tetrahydro-6,8,11-trihydroxy-8-[(1S)-1-hydroxyethyl]-1-methoxy-, hydrochloride, (8S,10S)- (9CI), L-lyxo-Hexopyranoside, 1,2,3,4,6,11-hexahydro-3,5,12-trihydroxy-3-(1-hydroxyethyl)-10-methoxy-6,11-dioxo-1-naphthacenyl 3-amino-2,3,6-trideoxy-, hydrochloride, (1S,3S)- α- (8CI), 13-Dihydrodaunomycin hydrochloride, 13-Dihydrodaunorubicin hydrochloride, Antibiotic 20798 RP, Antibiotic RP 20798, Dihydrodaunomycin hydrochloride, NSC 180510, RP 20798, (8S,10S)-10-[(3-Amino-2,3,6-trideoxy-α-L-lyxo-hexopyranosyl)oxy]-7,8,9,10-tetrahydro-6,8,11-trihydroxy-8-[(1S)-1-hydroxyethyl]-1-methoxy-5,12-naphthacenedione hydrochloride, Daunomycinol hydrochloride, Daunorubicinol hydrochloride, Duborimycin hydrochloride, Leukaemomycin D hydrochloride, (8S,10S)-10-[(3-Amino-2,3,6-trideoxy-α-L-lyxo-hexopyranosyl)oxy]-6,8,11-trihydroxy-8-[(1S)-1-hydroxyethyl]-1-methoxy-7,8,9,10-tetrahydronaphthacene-5,12-dione hydrochloride, (8S,10S)-10-[(3-Amino-2,3,6-trideoxy-α-L-lyxo-hexopyranosyl)oxy]-7,8,9,10-tetrahydro-6,8,11-trihydroxy-8-[1-hydroxyethyl]-1-methoxy-5,12-naphthacenedione Hydrochloride, 13-Dihydrodaunorubicin Hydrochloride
InChI Formula InChI=1S/C27H31NO10.ClH/c1-10-22(30)14(28)7-17(37-10)38-16-9-27(35,11(2)29)8-13-19(16)26(34)21-20(24(13)32)23(31)12-5-4-6-15(36-3)18(12)25(21)33;/h4-6,10-11,14,16-17,22,29-30,32,34-35H,7-9,28H2,1-3H3;1H/t10-,11-,14-,16-,17-,22+,27-;/m0./s1
SMILES Cl.COc1cccc2C(=O)c3c(O)c4C[C@](O)(C[C@H](O[C@H]5C[C@H](N)[C@H](O)[C@H](C)O5)c4c(O)c3C(=O)c12)[C@H](C)O
IUPAC Name (7S,9S)-7-[(2R,4S,5S,6S)-4-amino-5-hydroxy-6-methyloxan-2-yl]oxy-6,9,11-trihydroxy-9-[(1S)-1-hydroxyethyl]-4-methoxy-8,10-dihydro-7H-tetracene-5,12-dione;hydrochloride
Formula Weight 565.1715
Percent Purity >95%
Chemical Name or Material Daunorubicinol Hydrochloride (Mixture of Diastereomers)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.