Learn More
Daxx Antibody (CL3580) [DyLight 488], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-61622G
Product Code. 30287529
  • 737.00 € / 0.10mL
Description
Daxx Monoclonal antibody specifically detects Daxx in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Daxx
Monoclonal
DyLight 488
BING2, CENP-C binding protein, DAP6MGC126245, Daxx, death domain-associated protein 6, death-associated protein 6, death-domain associated protein, EAP1, ETS1-associated protein 1, Fas death domain-associated protein, Fas-binding protein, hDaxx, MGC126246
This antibody was developed using a recombinant protein derived from Q9UER7, with the exact immunogen sequence remaining proprietary.
0.1 mL
Apoptosis, Cancer, Phospho Specific, Tumor Suppressors
1616
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL3580
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified