missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DAZAP1 Antibody (SR1732), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22585-100ul
This item is not returnable.
View return policy
Description
DAZAP1 Monoclonal antibody specifically detects DAZAP1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| DAZAP1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 26528 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot | |
| SR1732 | |
| Western Blot 1:500-1:2000 | |
| DAZ associated protein 1, DAZ-associated protein 1, deleted in azoospermia associated protein 1, Deleted in azoospermia-associated protein 1, MGC19907 | |
| A synthesized peptide derived from human DAZAP1 (Uniprot #: Q96EP5) | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction