missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DAZAP2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
193.00 € - 463.00 €
Specifications
| Antigen | DAZAP2 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18632701
|
Novus Biologicals
NBP2-92144-0.02ml |
0.02 mL |
193.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18627800
|
Novus Biologicals
NBP2-92144-0.1ml |
0.1 mL |
463.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DAZAP2 Polyclonal antibody specifically detects DAZAP2 in Mouse samples. It is validated for Western BlotSpecifications
| DAZAP2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| DAZ associated protein 2, deleted in azoospermia associated protein 2, Deleted in azoospermia-associated protein 2, KIAA0058DAZ-associated protein 2, MGC14319, MGC766, proline-rich transcript in brain, proline-rich transcript, brain-expressed protein, PRTB | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-168 of human DAZAP2 (NP_055579.1). MNSKGQYPTQPTYPVQPPGNPVYPQTLHLPQAPPYTDAPPAYSELYRPSFVHPGAATVPTMSAAFPGASLYLPMAQSVAVGPLGSTIPMAYYPVGPIYPPGSTVLVEGGYDAGARFGAGATAGNIPPPPPGCPPNAAQLAVMQGANVLVTQRKGNFFMGGSDGGYTIW | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 9802 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title