missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DC-LAMP Rabbit anti-Human, Clone: 2, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89551-100UL
This item is not returnable.
View return policy
Description
DC-LAMP Monoclonal specifically detects DC-LAMP in Human samples. It is validated for ELISA, Sandwich ELISA Detection.
Specifications
| DC-LAMP | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 2 | |
| ELISA 1:5000-1:10000, Sandwich ELISA Detection 1:1000-1:10000 | |
| CD208 antigen, DC-LAMP, DCLAMPDC-lysosome-associated membrane glycoprotein, LAMP, LAMP-3, lysosomal-associated membrane protein 3DC LAMP, lysosome-associated membrane glycoprotein 3, Protein TSC403, TSC403CD208 | |
| Recombinant Human DC-LAMP protein | |
| 100 μg | |
| Dendritic Cell Markers | |
| 27074 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction