missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DC-SIGN/CD209 Antibody (C209/1781), Novus Biologicals™
Mouse Monoclonal Antibody
299.00 € - 555.00 €
Specifications
| Antigen | DC-SIGN/CD209 |
|---|---|
| Clone | C209/1781 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, SDS-Page |
| Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), SDS-Page |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18057503
|
Novus Biologicals
NBP2-53275-20UG |
20 μg |
299.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18188509
|
Novus Biologicals
NBP2-53275-100UG |
100 μg |
555.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DC-SIGN/CD209 Monoclonal specifically detects DC-SIGN/CD209 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| DC-SIGN/CD209 | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), SDS-Page | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Immunology, Virology Bacteria and Parasites | |
| 10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| 30835 | |
| Recombinant human CD209 protein fragment | |
| Primary | |
| Store at 4C. |
| C209/1781 | |
| Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, SDS-Page | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD209 antigendendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbingnon-integrin, CD209 molecule, CDSIGNHIV gpl20-binding protein, CLEC4LC-type lectin domain family 4 member L, DC-SIGN1C-type lectin domain family 4, member L, DC-SIGNMGC129965, Dendritic cell-specific ICAM-3-grabbing non-integrin 1 | |
| CD209 | |
| IgG2b κ | |
| Protein A or G purified | |
| 45 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title