missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCDC2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-38184
This item is not returnable.
View return policy
Description
DCDC2 Polyclonal specifically detects DCDC2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| DCDC2 | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Q9UHG0 | |
| DCDC2 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: DAPEQVEEILDHSEQQARPARVNGGTDEENGEELQQVNNELQLVLDKERKSQGAGSGQDEADVDPQRPPRPEVKITSPEENENNQQNKDYAAVA | |
| 0.1 mL | |
| Signal Transduction, Vision | |
| 51473 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| DCDC2A, doublecortin domain containing 2, KIAA1154Protein RU2S, RU2doublecortin domain-containing protein 2, RU2S | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction