missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCDC2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-38184-25ul
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
DCDC2 Polyclonal specifically detects DCDC2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Especificaciones
| DCDC2 | |
| Polyclonal | |
| Western Blot 0.4 μg/mL, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Q9UHG0 | |
| DCDC2 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: DAPEQVEEILDHSEQQARPARVNGGTDEENGEELQQVNNELQLVLDKERKSQGAGSGQDEADVDPQRPPRPEVKITSPEENENNQQNKDYAAVA | |
| 25 μL | |
| Signal Transduction, Vision | |
| 51473 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| DCDC2A, doublecortin domain containing 2, KIAA1154Protein RU2S, RU2doublecortin domain-containing protein 2, RU2S | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido