missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DCL-1 Monoclonal Antibody (662714)
Rat Monoclonal Antibody
Brand: Invitrogen™ MA561665
This item is not returnable.
View return policy
Description
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
CD302 is a C-type lectin receptor involved in cell adhesion and migration, as well as endocytosis and phagocytosis.
Specifications
| DCL-1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9DCG2 | |
| Cd302 | |
| Mouse myeloma cell line NS0-derived recombinant mouse CD302/CLEC13A, Met1-Cys165. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 662714 | |
| Unconjugated | |
| Cd302 | |
| 1110055L24Rik; AI159627; BIMLEC; CD302; CD302 antigen; CD302 molecule; CLEC13A; C-type lectin BIMLEC; C-type lectin domain family 13 member A; C-type lectin domain family 13, member A; DCL1; DCL-1; DEC205-associated C-type lectin 1; KIAA0022; RGD1307606; Type I transmembrane C-type lectin receptor DCL-1 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 66205 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction