missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCLK1 Antibody (S05-8D6), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19952-100ul
This item is not returnable.
View return policy
Description
DCLK1 Monoclonal antibody specifically detects DCLK1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| DCLK1 | |
| Monoclonal | |
| 0.3 mg/mL | |
| Western Blot, Immunohistochemistry 1:20, Immunohistochemistry-Paraffin | |
| DCAMKL1CLICK1, DCDC3ACL1, DCLK, doublecortin and CaM kinase-like 1, Doublecortin domain-containing protein 3A, doublecortin-like kinase 1Doublecortin-like and CAM kinase-like 1, EC 2.7.11, KIAA0369EC 2.7.11.1, serine/threonine-protein kinase DCLK1 | |
| A synthetic peptide of human DCLK1 (Uniprot # O15075) | |
| 100 μg | |
| Protein Kinase | |
| 9201 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| S05-8D6 | |
| Unconjugated | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction