missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14995-1-AP-20UL
This item is not returnable.
View return policy
Description
May be involved in ubiquitination and subsequent proteasomal degradation of target proteins. Component of the DDD- E2 complexes which may provide a platform for interaction with CUL4A and WD repeat proteins.Specifications
| DDA1 | |
| Polyclonal | |
| Unconjugated | |
| DDA1 | |
| C19orf58, DDA1, DET1 and DDB1 associated 1, PCIA 1, PCIA1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 66498, 688813, 79016 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunocytochemistry, Western Blot, Immunofluorescence | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9BW61, Q9D9Z5 | |
| Dda1 | |
| DDA1 Fusion Protein Ag6981 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction