missing translation for 'onlineSavingsMsg'
Learn More

DDR Inhibitor, MedChemExpress

Product Code. 16432698
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16432698

Brand: Medchem Express HYW018931

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress DDR Inhibitor is a potent discoidin domain receptor (DDR) inhibitor, with an IC50 of 3.3 nM for DDR2, and shows 53% inhibition on DDR1 at 1.5 nM.

DDR Inhibitor is a potent discoidin domain receptor (DDR) inhibitor, with an IC50 of 3.3 nM for DDR2, and shows 53% inhibition on DDR1 at 1.5 nM.

Specifications

Chemical Name or Material DDR Inhibitor
CAS 1644069-80-6
Color White
Purity Grade Notes Research
Molecular Formula C23H20FN5O2
Quantity 5 mg
Solubility Information DMSO : 65 mg/mL (155.71 mM; Need ultrasonic)
SMILES O=C(C1=CN=C2C=CC=CN21)NC3=CC(CNC(NC4=CC=CC(F)=C4)=O)=CC=C3C
Molecular Weight (g/mol) 417.44
Formula Weight 417.44
Percent Purity 99.43%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.