missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX17 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
198.00 € - 462.00 €
Specifications
| Antigen | DDX17 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
DDX17 Polyclonal antibody specifically detects DDX17 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| DDX17 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Translation Control, Vision | |
| PBS with 50% glycerol, pH7.3. | |
| 10521 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| DEAD (Asp-Glu-Ala-Asp) box polypeptide 17, DEAD box protein 17, DEAD box protein p72, DEAD/H box 17, DKFZp761H2016, EC 3.6.1, EC 3.6.4.13, P72DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 17 (72kD), probable ATP-dependent RNA helicase DDX17, RH70, RNA-dependent helicase p72 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 170-240 of human DDX17 (NP_006377.2). VFAFHHANFPQYVMDVLMDQHFTEPTPIQCQGFPLALSGRDMVGIAQTGSGKTLAYLLPAIVHINHQPYLE | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title