Learn More
DDX21 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NB100-1718MFV450
Product Code. 18741183
  • 629.00 € / 0.10mL
Description
DDX21 Polyclonal antibody specifically detects DDX21 in Human, Mouse, Zebrafish samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DDX21
Flow Cytometry
Flow Cytometry
Store at 4C in the dark.