missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DDX21 Recombinant Rabbit Monoclonal Antibody (K01_1Q31)
GREENER_CHOICE

Product Code. 30824654
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30824654 100 μL
30830427 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30824654 Supplier Invitrogen™ Supplier No. MA563520

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: LHGELQDRKR GRA.

Eukaryotic initiation factor 2B (EIF2B), which is necessary for protein synthesis, is a GTP exchange factor composed of five different subunits. The protein encoded by this gene is the fourth, or delta, subunit. Defects in this gene are a cause of leukoencephalopathy with vanishing white matter (VWM) and ovarioleukodystrophy. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDX21
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone K01_1Q31
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 150mM NaCl, 0.05% BSA, 40% glycerol and 0.01% sodium azide; pH 7.4
Gene DDX21
Gene Accession No. Q9NR30
Gene Alias AI255159; AL022742; D10Ertd645e; D10Wsu42e; DDX21; Ddx21a; DEAD (Asp-Glu-Ala-Asp) box helicase 21; DEAD (Asp-Glu-Ala-Asp) box polypeptide 21; DEAD (Asp-Glu-Ala-Asp) box polypeptide 21a; DEAD box protein 21; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 21; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 21 (RNA helicase II/Gu); DEAD-box helicase 21; DExD-box helicase 21; Gu protein; GUA; gu-alpha; GURDB; Nucleolar RNA helicase 2; nucleolar RNA helicase Gu; Nucleolar RNA helicase II; RH II/Gu; RH-II/GU; RH-II/GuA; RH-II/Gualpha; RNA helicase II/Gu alpha
Gene Symbols DDX21
Host Species Rabbit
Immunogen A synthetic peptide of human DDX21 (aa 248-260).
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9188
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.