missing translation for 'onlineSavingsMsg'
Learn More

DDX4 Monoclonal Antibody (OTI1D11), TrueMAB™, OriGene

Product Code. 17568224
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17568224 30 μL 30µL
17548224 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17568224 Supplier Origene Technologies Supplier No. TA813367S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DDX4 Monoclonal antibody specifically detects DDX4 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen DDX4
Applications Western Blot
Classification Monoclonal
Clone OTI1D11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene DDX4
Gene Accession No. Q61496, Q64060, Q9NQI0
Gene Alias ATP-dependent RNA helicase DDX4; AV206478; Cvh; Ddx4; D-E-A-D (aspartate-glutamate-alanine-aspartate) box polypeptide 4; DEAD (aspartate-glutamate-alanine-aspartate) box polypeptide 4; DEAD (Asp-Glu-Ala-Asp) box polypeptide 4; DEAD box polypeptide 4; DEAD box protein 4; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 4; DEAD-box helicase 4; MGC111074; Mvh; mvh / m'vasa; probable ATP-dependent RNA helicase DDX4; putative ATP-dependent RNA helicase DDX4; RVLG; VASA; Vasa homolog; vasa-like gene protein
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-400 of human DDX4 (NP_077726) produced in E.coli.
Purification Method Protein A/G
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13206, 310090, 54514
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.