missing translation for 'onlineSavingsMsg'
Learn More

Decafluorobiphenyl, SPEX CertiPrep™

Product Code. 12969144 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12969144 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12969144 Supplier SPEX Certiprep Supplier No. S1103

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Regulatorischer Hinweis: Abgabe sachkundepflichtiger Stoffe und Gemische nur bei Erfüllung der Vorrausetzungen nach Chemikalienverbotsverordnung (ChemVerbotsV). Keine Abgabe an Privatpersonen.

Beinhaltet

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Decafluorobiphenyl434-90-21000μg/mL

Zertifizierungen

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Spezifikation

Packaging 2mL amber prescored glass ampule
CAS 434-90-2, 67-56-1
CAS Min % 0.1, 99.9
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
CAS Max % 0.1, 99.9
Methods For US EPA Method 551, 625
Molecular Formula C12F10
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
InChI Key ONUFSRWQCKNVSL-UHFFFAOYSA-N
SMILES C1(=C(C(=C(C(=C1F)F)F)F)F)C2=C(C(=C(C(=C2F)F)F)F)F
IUPAC Name 1,2,3,4,5-pentafluoro-6-(2,3,4,5,6-pentafluorophenyl)benzene
Solvent or Matrix Methanol
Molecular Weight (g/mol) 334.116
PubChem CID 67949
Quantity 1 mL
Chemical Name or Material Decafluorobiphenyl
Mehr anzeigen Weniger anzeigen
compliance-icons compliance-icons compliance-icons
Produktidentifikator
  • Decafluorobiphenyl
Signalwort
  • Danger
Gefahrenkategorie
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
Gefahrenhinweis
  • H225-Highly flammable liquid and vapour.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Sicherheitshinweis
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Ergänzende Informationen
  • MIXTURE LIST-Contain: methanol

Product intended for laboratory use only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.