missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Decapentaplegic Polyclonal Antibody
GREENER_CHOICE

Product Code. 30534576
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30534576 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30534576 Supplier Invitrogen™ Supplier No. PA5148509

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chicken Polyclonal Antibody

Reconstitute at 0.2 mg/mL in sterile PBS.

Decapentaplegic (Dpp) is a gene in Drosophila melanogaster, encoding a protein that is a member of the Transforming Growth Factor-beta (TGF-beta) superfamily. Dpp plays a crucial role in various developmental processes, including embryogenesis, organogenesis, and the regulation of cell growth and differentiation. It functions primarily as a morphogen, a signaling molecule that forms a concentration gradient to provide positional information to cells, guiding their development and differentiation. Dpp is essential for the proper patterning of tissues and organs, such as the wings, legs, and gut in fruit flies. Mutations in the Dpp gene can lead to severe developmental abnormalities and defects in tissue patterning. The study of Dpp and its signaling pathways provides valuable insights into the mechanisms of morphogen-mediated development and has broader implications for understanding similar TGF-beta signaling pathways in other organisms, including humans.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Decapentaplegic
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with trehalose and no preservative
Host Species Chicken
Immunogen E. coli-derived recombinant Drosophila Decapentaplegic/DPP, Asp457-Arg588 (Gln473His, Pro474Ala).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Drosophila
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.