Learn More
Decorin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14667-1-AP-20UL
 View more versions of this product
Product Code. 16823424
  • 145.00 € / 20µL
Description
Decorin is a member of the small, leucine-rich proteoglycan (SLRP) family along with biglycan. Found in the interterritorial region of cartilage, decorin interacts with collagen and other matrix proteins to regulate the cartilaginous matrix. It has been shown to interact with fibronectin, thrombospondin, C1q, epidermal growth factor receptor (EGFR), and TGF-b. It is believed that decorin also plays a role in the cell cycle of chondrocytes. The decorin neoepitope fragment has a sequence of PKTLQE, and is generated by MMP-3 or ADAMTS-4 cleavage at Glu154-Leu155 of decorin core protein.
Specifications
Specifications
Decorin
Polyclonal
Unconjugated
DCN
Bone proteoglycan II, CSCD, DCN, Decorin, DSPG2, PG S2, PG40, PGII, PGS2, SLRR1B
Rabbit
Antigen Affinity Chromatography
RUO
13179, 1634, 29139
-20°C
Liquid
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.18 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P07585, P28654, Q01129
Dcn
Decorin Fusion Protein Ag6328
20 μL
Primary
Rat, Mouse, Human