missing translation for 'onlineSavingsMsg'
Learn More

Decyl decanoate, 98%

Product Code. 15525946 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15525946 25 mL 25mL
15535946 100 mL 100mL
15545946 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15525946 Supplier Thermo Scientific Acros Supplier No. 457110250
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 1654-86-0 | C20H40O2 | 312.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1654-86-0
Molecular Formula C20H40O2
Molecular Weight (g/mol) 312.54
InChI Key XAKXZZPEUKNHMA-UHFFFAOYSA-N
Synonym decanoic acid, decyl ester, decanoic acid decyl ester, unii-b88b7acl5l, b88b7acl5l, n-capric acid n-decyl ester, capryl caprate, acmc-209dsk, decanoic acid, decylester
PubChem CID 74247
IUPAC Name decyl decanoate
SMILES CCCCCCCCCCOC(=O)CCCCCCCCC

Specifications

Density 0.8590g/mL
Boiling Point 174.0°C (2.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Linear Formula CH3(CH2)8CO2(CH2)9CH3
Quantity 25 mL
Specific Gravity 0.859
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol
Formula Weight 312.54
Percent Purity 98%
Chemical Name or Material Decyl decanoate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.