Learn More
Fisherbrand™ Dedicated Platform
Product Code. 16373246 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Holds:
12 x 125 mL Flasks
4 Microplates
5 x 500 mL Flasks
8 x 250 mL mL Flasks
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Unit Size
16373246 4 Microplates Each
16363246 5 x 500 mL Flasks Each
16353246 8 x 250 mL mL Flasks Each
16343246 12 x 125 mL Flasks Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16373246
  • ONLINE EXCLUSIVE PRICE
    678.00 €
    581.88 € / Pack of 1
    Save 96.12 €
    14% Off
Estimated Shipment: 02-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Fisherbrand™ Dedicated Platform is designed for use with a shaker.
  • Non-slip
  • Rubber-coated surface
  • For use with shaker
Specifications
Holds 4 Microplates
Product Type Dedicated Platform
Material Aluminum, Stainless Steel
For Use With (Equipment) Mini Shaking Incubator
No. of Positions 4