missing translation for 'onlineSavingsMsg'
Learn More

DEET, TRC

Product Code. 30135137
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
100g
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135137 100 g 100g
30011664 10 g 10g
17953115 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135137 Supplier Toronto Research Chemicals Supplier No. TRCD228500100G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134-62-3
Molecular Formula C12 H17 N O
Molecular Weight (g/mol) 191.27
Synonym m-Toluamide, N,N-diethyl- (6CI,7CI,8CI), N,N-Diethyl-3-methylbenzamide, 3-Methyl-N,N-diethylbenzamide, AI 3-22542, Amincene C 140, Amincene C-EM, Amway Hour Guard, Autan, Bepper DET, Cutter Unscented, DEET, DET, DET (insect repellent), DETA, Delphene, Detamide, Dieltamid, Diethyl-m-toluamide, Diethyltoluamide, ENT 20218, ENT 22542, Finish MIT, Flypel, M-Det, Metadelfene, Metadelphene, N,N-Diethyl-3-methylbenzoylamide, N,N-Diethyl-m-methylbenzamide, N,N-Diethyl-m-toluamide, N,N-Diethyl-m-toluamide, N,N-Diethyl-m-tolylamide, N,N-Diethyl-meta-toluamide, N,N'-Diethyl-3-methylbenzamide, NSC 33840, Naugatuck DET, Off, Off! Skintastic, Repper DET, Repudin-Special, Sawyer Control Release Formula, Sawyer Controlled-Release, Ultra 30, Ultrathon, Vaseline Mosquito Repellent, m-DETA, m-Delphene, m-Toluic acid diethylamide
IUPAC Name N,N-diethyl-3-methylbenzamide
SMILES CCN(CC)C(=O)c1cccc(C)c1

Specifications

Quantity 100 g
InChI Formula InChI=1S/C12H17NO/c1-4-13(5-2)12(14)11-8-6-7-10(3)9-11/h6-9H,4-5H2,1-3H3
Formula Weight 191.131
Percent Purity >95%
Chemical Name or Material Diethyltoluamide (DEET)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.