Learn More
OHAUS™ Defender™ 3000 - I-D33 Bench Scale, Certified
Bench Scales for Basic Industrial Applications
Brand:  OHAUS™ 30685116
Additional Details : Weight : 7.70900kg Transport : UN number : 2800 Chem class : 8
 View more versions of this product
Product Code. 17562146
  • 575.00 € / Each
Estimated Shipment: 14-05-2024
to see stock.
Description
Description
  • Results are clearly shown using the large, bright backlit LCD display.
  • In Checkweighing mode display changes color (yellow/green/red) to alert when within the target weight range.
  • This industrial scale features a 304 stainless steel platform, powder-coated steel tubular frame, and aluminum load cell.
  • Indicator features a stainless steel housing and mounting bracket.
  • Features software for Parts Counting, Checkweighing and Totalization.
  • Includes RS232 with optional USB Device or Ethernet for printing or data connectivity.
Specifications
Specifications
100 Hours with Rechargeable Battery;360 Hours with Disposable Batteries
Standard RS232 and optional USB Device or Ethernet
15 kg
355 x 305 mm
D33P15B1R5
Weighing, Checkweighing, Counting, Totalization.
10.2 kg
6 C (LR14) Batteries (Not Included);AC Adapter (Included);Rechargeable Battery (Not Included)
ABS plastic indicator, 304-stainless steel platform, powder-coated steel tubular frame, aluminum load cell, non-slip adjustable rubber feet
7 Segment LCD with Multicolor Backlight
5 g
-10°C to +40°C, 85%RH, non-condensing
646 x 305 x 130 mm