Learn More
Description
Defined Keratinocyte SFM is a serum-free defined medium optimized for the isolation and expansion of human keratinocytes without the need for bovine pituitary extract (BPE) supplementation or the use of fibroblast feeder layers.
The growth-promoting activity of BPE has been replaced by the inclusion of growth promoting agents which enhance consistent product performance. These growth-promoting agents also maintain cell morphology, physiological markers, and growth. The medium is formulated to inhibit fibroblast proliferation and has a low calcium concentration (<0.1 mM), allowing for the isolation and culture of highly enriched populations of undifferentiated keratinocytes.
Note: This formulation contains some human and animal origin components.
Note: A Defined Keratinocyte-SFM Master File has been submitted to the FDA. Permission to cross-reference the Master File may be obtained by contacting Technical Services or your local Sales Representative. Defined Keratinocyte-SFM consists of a basal liquid medium and frozen supplement which is added at the time of use. The fully supplemented medium has a shelf-life of 10 weeks under normal use conditions. Product performance is confirmed by evaluation of human primary keratinocyte growth.
Specifications
Specifications
| Cell Type | Keratinocyte Cells, Epithelial Cells |
| Classification | Serum-free |
| Concentration | 1 X |
| Form | Liquid |
| Product Type | Kit |
| Serum Level | Serum-free |
| Sterility | Sterile-filtered |
| Sterilization Method | Sterile-filtered |
| With Additives | Glutamine, Phenol Red |
| Quantity | 500 mL |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
