missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DEGS1 Recombinant Rabbit Monoclonal Antibody (K01_1T94)

Product Code. 30830444
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30830444 50 μL
30828433 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30830444 Supplier Invitrogen™ Supplier No. MA564902
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

DEGS1 Recombinant Monoclonal Antibody for Western Blot

The product of the DEGS1 gene is a member of the membrane fatty acid (sphingolipid delta 4 ) desaturase family. Reported to function by inserting double bonds at delta 4 position in fatty acids. DEGS1 is an intracellular enzyme. Overexpression of this gene inhibited biosynthesis of the EGF receptor, suggesting a possible role of a fatty acid desaturase in regulation biosynthetic processing of the EGF receptor. Two splice variants have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DEGS1
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1T94
Concentration 1.30 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 150mM NaCl, 40% glycerol and 0.01% sodium azide; pH 7.4
Gene DEGS1
Gene Accession No. O15121
Gene Alias AA536663; Cell migration-inducing gene 15 protein; Degenerative spermatocyte homolog 1; degenerative spermatocyte homolog 1 (Drosophila); degenerative spermatocyte homolog 1, lipid desaturase; degenerative spermatocyte homolog, lipid desaturase; degenerative spermatocyte-like protein RDES; Degs; DEGS1; DEGS-1; delta 4-desaturase, sphingolipid 1; Delta(4)-desaturase; delta(4)-desaturase, sphingolipid 1; DES1; Des-1; dihydroceramide desaturase; dihydroceramide desaturase 1; dihydroceramide desaturase-1; DSH1; FADS7; fb81d06; hypothetical protein LOC507290; im:6909319; Mdes; membrane fatty acid (lipid) desaturase; membrane lipid desaturase; MGC5079; MIG15; migration-inducing gene 15 protein; MLD; sphingolipid 1; sphingolipid delta 4 desaturase; sphingolipid delta(4)-desaturase 1; sphingolipid delta(4)-desaturase DES1; wu:fa25h01; wu:fb81d06
Gene Symbols DEGS1
Host Species Rabbit
Immunogen A synthetic peptide of human MLD.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8560
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.