missing translation for 'onlineSavingsMsg'
Learn More

(+)-Dehydroabietylamine hydrochloride

Product Code. 15465389
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15465389 10 mg 10mg
15475389 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15465389 Supplier Thermo Scientific Alfa Aesar Supplier No. J63123.MA
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 16496-99-4 | C20H32ClN | 321.93 g/mol

Resolving agent for carboxylic acids

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16496-99-4
Molecular Formula C20H32ClN
Molecular Weight (g/mol) 321.93
MDL Number MFCD06795849
InChI Key CVPQLGCAWUAYPF-UHFFFAOYNA-N
Synonym leelamine hydrochloride, dehydroabiethylamine hydrochloride, +-dehydroabietylamine, lylamine hydrochloride, nsc 2955 hydrochloride, +-dehydroabietylamine hydrochloride, +-dehydroabiethylamine hydrochloride, abieta-8,11,13-trien-18-amine-hydrogen chloride 1/1, 13-isopropyl-podocarpa-8,11,13-trien-15-amine hydrochloride, 1r,4as,10ar-7-isopropyl-1,4a-dimethyl-1,2,3,4,4a,9,10,10a-octahydrophenanthren-1-yl methanamine hydrochloride
PubChem CID 16759156
IUPAC Name [(1R,4aS,10aR)-1,4a-dimethyl-7-propan-2-yl-2,3,4,9,10,10a-hexahydrophenanthren-1-yl]methanamine;hydrochloride
SMILES Cl.CC(C)C1=CC2=C(C=C1)C1(C)CCCC(C)(CN)C1CC2

Specifications

Packaging Plastic bottle
Quantity 10 mg
Beilstein 3084620
Formula Weight 321.9
Physical Form Solid
Chemical Name or Material (+)-Dehydroabietylamine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.