missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Delta-hemolysin Polyclonal Antibody
GREENER_CHOICE

Product Code. 17197458 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
400µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17197458 400 μL 400µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17197458 Supplier Invitrogen™ Supplier No. PA5116227

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Delta-hemolysin Polyclonal Antibody for Western Blot

Bacterial toxin that lyses erythrocytes and many other mammalian cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Delta-hemolysin
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene hld
Gene Accession No. P0C1V1
Gene Symbols hld
Host Species Rabbit
Immunogen KLH conjugated peptide 1-30aa from delta hemolysin protein of staphylococcus aureus.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16042116
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.