missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Deltamethrin 1000 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Deltamethrin |
| CAS | 52918-63-5 |
| Analyte or Component Names | Deltamethrin |
| Shipping Condition | +20°C |
| Molecular Formula | C22 H19 Br2 N O3 |
| Concentration | 1000 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | (1R,3R)-3-(2,2-Dibromoethenyl)-2,2-dimethylcyclopropanecarboxylic acid (S)-cyano(3-phenoxyphenyl)methyl ester, [1R-[1α(S*),3α]]-3-(2,2-Dibromoethenyl)-2,2-dimethylcyclopropanecarboxylic acid cyano(3-phenoxyphenyl)methyl ester, (S)-Cyano(3-phenoxyphenyl)methyl (1R,3R)-3-(2,2-dibromoethenyl)-2,2-dimethylcyclopropanecarboxylate, Alpha Max, AquaK'othrine, Battalion, Butox, Cislin, Crackdown, Decamethrin, Decamethrine, Decis, Decis 0.5ULV, Decis 1.5ULV, Decis 2.5ULV, Decis Extra, Decis Super, Decis flow 2.5, Decistab, Decistab TB, Delta Gold, DeltaGard, Deltacid, Deltadust, Deltagran, Deltamethrin, Ezeetab, FMC 45498, Glossinex 200, IPO 8831, K-O Tab, K-Obiol, K-Obiol 2P, K-Othrin, K-Othrine, NRDC 161, New Musigie, Pendelta, Phagase 1, RU 22974, RUP 987, Scalibor, Suspend, Timbertreat DM 5, Ultratec, Vectocid, Zorcis |
| InChI Formula | InChI=1S/C22H19Br2NO3/c1-22(2)17(12-19(23)24)20(22)21(26)28-18(13-25)14-7-6-10-16(11-14)27-15-8-4-3-5-9-15/h3-12,17-18,20H,1-2H3/t17-,18+,20-/m0/s1 |
| SMILES | CC1(C)[C@@H](C=C(Br)Br)[C@H]1C(=O)O[C@H](C#N)c2cccc(Oc3ccccc3)c2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?