missing translation for 'onlineSavingsMsg'
Learn More

Demeclocycline hydrochloride, MedChemExpress

Product Code. 16497366
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16497366 100 mg 100mg
16481876 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16497366 Supplier Medchem Express Supplier No. HY17560
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Demeclocycline hydrochloride is a tetracycline antibiotic; is an antibiotic in the treatment of Lyme disease, acne, and bronchitis.

Demeclocycline hydrochloride is a tetracycline antibiotic

Specifications

Chemical Name or Material Demeclocycline hydrochloride
CAS 64-73-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C21H22Cl2N2O8
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (199.48 mM; Need ultrasonic) ∣H2O : 20 mg/mL (39.90 mM; Need ultrasonic and warming)
SMILES O=C(C(C1=O)=C(O)[C@@H](N(C)C)[C@]2([H])C[C@]3([H])[C@H](O)C4=C(C(C3=C(O)[C@@]21O)=O)C(O)=CC=C4Cl)N.Cl
Molecular Weight (g/mol) 501.31
Formula Weight 501.31
Percent Purity 95.09%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.