missing translation for 'onlineSavingsMsg'
Learn More

Demethoxycurcumin, MedChemExpress

Product Code. 16448818
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16448818 10 mg 10mg
16421268 5 mg 5mg
16404727 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16448818 Supplier Medchem Express Supplier No. HYN0006
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Demethoxycurcumin(Curcumin II) is a major active curcuminoid; possess anti-inflammatory properties; also exert cytotoxic effects in human cancer cells via induction of apoptosis.

Demethoxycurcumin(Curcumin II) is a major active curcuminoid

Specifications

Chemical Name or Material Demethoxycurcumin
CAS 22608-11-3
Color Red
Purity Grade Notes Research
Molecular Formula C20H18O5
Quantity 10 mg
Synonym Curcumin II Desmethoxycurcumin Monodemethoxycurcumin
Solubility Information DMSO : 100 mg/mL (295.55 mM; Need ultrasonic)
SMILES O=C(CC(/C=C/C1=CC=C(O)C=C1)=O)/C=C/C2=CC=C(O)C(OC)=C2.[(E)].[(E)]
Molecular Weight (g/mol) 338.35
Formula Weight 338.35
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.