missing translation for 'onlineSavingsMsg'
Learn More

Demeton-O, TRC

Product Code. 30040415
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30040415 100 mg 100mg
18082550 25 mg 25mg
18001060 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30040415 Supplier Toronto Research Chemicals Supplier No. TRCD231498100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 298-03-3
Molecular Formula C8 H19 O3 P S2
Molecular Weight (g/mol) 258.34
Synonym S-[2-(Ethylthio)ethyl] O,O-dimethylphosphorothioate, Demetone-O
IUPAC Name diethoxy-(2-ethylsulfanylethoxy)-sulfanylidene-λ^{5}-phosphane
SMILES CCOP(=S)(OCC)OCCSCC

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C8H19O3PS2/c1-4-9-12(13,10-5-2)11-7-8-14-6-3/h4-8H2,1-3H3
Formula Weight 258.0513
Percent Purity >95%
Chemical Name or Material Demeton-O
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.