missing translation for 'onlineSavingsMsg'
Learn More

Demeton-S Sulfone, TRC

Product Code. 30137072
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137072 100 mg 100mg
18011940 5 mg 5mg
18072850 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137072 Supplier Toronto Research Chemicals Supplier No. TRCD231385100MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2496-91-5
Molecular Formula C8 H19 O5 P S2
Molecular Weight (g/mol) 290.34
Synonym Ethanethiol, 2-(ethylsulfonyl)-, S-ester with O,O-diethyl phosphorothioate (8CI), Demeton sulfone, Demeton thiol sulfone, Demeton-S sulfone, Isosystox sulfone, O,O-Diethyl S-[2-(ethylsulfonyl)ethyl] phosphorothioate
IUPAC Name 1-diethoxyphosphorylsulfanyl-2-ethylsulfonylethane
SMILES CCOP(=O)(OCC)SCCS(=O)(=O)CC

Specifications

Analyte or Component Names Disulfoton-oxon-sulfon
Shipping Condition Room Temperature
InChI Formula InChI=1S/C8H19O5PS2/c1-4-12-14(9,13-5-2)15-7-8-16(10,11)6-3/h4-8H2,1-3H3
Solution Type Neat
Industry Type Pharmaceutical
Quantity 100 mg
Formula Weight 290.0412
Chemical Name or Material Disulfoton-oxon-sulfon
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.