missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dengue Virus 2 Envelope Antibody (144) - (Domain III), Novus Biologicals™
Mouse Monoclonal Antibody
241.00 € - 448.00 €
Specifications
| Antigen | Dengue Virus 2 Envelope |
|---|---|
| Clone | 144 |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:1000-1:2000 |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18678268
|
Novus Biologicals
NBP3-05828-100ul |
100 μg |
448.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18636676
|
Novus Biologicals
NBP3-05828-50ul |
50 μg |
241.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dengue Virus 2 Envelope Monoclonal antibody specifically detects Dengue Virus 2 Envelope in Virus samples. It is validated for Western Blot, ELISASpecifications
| Dengue Virus 2 Envelope | |
| Western Blot 1:500-1:2000, ELISA 1:1000-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Virus | |
| Dengue 2 envelope;Dengue envelope;envelope protein, Envelope protein E | |
| IgG1 | |
| Protein A purified |
| 144 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites | |
| 0.2 um filtered solution in PBS | |
| Produced in mouse immunized with recombinant Dengue Virus 2 Envelope (Accession#: AAC59274.1; Met578-Gln680). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title