Learn More
Dengue Virus 2 Envelope Antibody (144), FITC , Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-05828F
Product Code. 18614826
  • 565.00 € / 0.10mL
Description
Dengue Virus 2 Envelope Monoclonal antibody specifically detects Dengue Virus 2 Envelope in Virus samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Dengue Virus 2 Envelope
Monoclonal
FITC
Envelope protein E
Produced in mouse immunized with recombinant Dengue virus (DENV) (type 2, strain New Guinea C/PUO-218 hybrid) E/ Envelope Protein (Domain III) (AAC59274.1; Met578-Gln680).
0.1 mL
Primary
Store at 4°C in the dark.
IgG
ELISA
144
PBS
Mouse
Protein A purified
RUO
Virus
Purified